MilliporeSigma
  • A nanodiamond/CNT-SiC monolith as a novel metal free catalyst for ethylbenzene direct dehydrogenation to styrene.
A nanodiamond/CNT-SiC monolith as a novel metal free catalyst for ethylbenzene direct dehydrogenation to styrene.
Chemical communications (Cambridge, England) (2014-06-07)
Hongyang Liu, Jiangyong Diao, Qi Wang, Songyuan Gu, Tong Chen, Changxi Miao, Weimin Yang, Dangsheng Su
ABSTRACT
A novel nanodiamond/CNT-SiC monolith catalyst has been prepared by a facile two-step approach. The as-synthesized monolith afforded high activity and stability for ethylbenzene direct dehydrogenation to styrene, showing its potential application as a metal free catalyst in gaseous catalytic reactions.
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%