MilliporeSigma
  • Single chain epidermal growth factor receptor antibody conjugated nanoparticles for in vivo tumor targeting and imaging.
Single chain epidermal growth factor receptor antibody conjugated nanoparticles for in vivo tumor targeting and imaging.
Small (Weinheim an der Bergstrasse, Germany) (2008-12-18)
Lily Yang, Hui Mao, Y Andrew Wang, Zehong Cao, Xianghong Peng, Xiaoxia Wang, Hongwei Duan, Chunchun Ni, Qingan Yuan, Gregory Adams, Mark Q Smith, William C Wood, Xiaohu Gao, Shuming Nie
ABSTRACT
Epidermal growth factor receptor (EGFR) targeted nanoparticle are developed by conjugating a single-chain anti-EGFR antibody (ScFvEGFR) to surface functionalized quantum dots (QDs) or magnetic iron oxide (IO) nanoparticles. The results show that ScFvEGFR can be successfully conjugated to the nanoparticles, resulting in compact ScFvEGFR nanoparticles that specifically bind to and are internalized by EGFR-expressing cancer cells, thereby producing a fluorescent signal or magnetic resonance imaging (MRI) contrast. In vivo tumor targeting and uptake of the nanoparticles in human cancer cells is demonstrated after systemic delivery of ScFvEGFR-QDs or ScFvEGFR-IO nanoparticles into an orthotopic pancreatic cancer model. Therefore, ScFvEGFR nanoparticles have potential to be used as a molecular-targeted in vivo tumor imaging agent. Efficient internalization of ScFvEGFR nanoparticles into tumor cells after systemic delivery suggests that the EGFR-targeted nanoparticles can also be used for the targeted delivery of therapeutic agents.
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 540 nm, 4 μM in 10 mM PBS
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, amine functionalized, fluorescence λem 600 nm, 4 μM in 10 mM PBS
Sigma-Aldrich
CdSe/ZnS core-shell type quantum dots, carboxylic acid functionalized, fluorescence λem 645 nm, 1 mg/mL in H2O