MilliporeSigma
  • Regulation of Cu-Zn superoxide dismutase on SCN2A in SH-SY5Y cells as a potential therapy for temporal lobe epilepsy.
Regulation of Cu-Zn superoxide dismutase on SCN2A in SH-SY5Y cells as a potential therapy for temporal lobe epilepsy.
Molecular medicine reports (2013-11-14)
Jun Xiang, Yugang Jiang
ABSTRACT
In order to evaluate SCN2A as a candidate gene for epileptic susceptibility and the use of a Cu-Zn superoxide dismutase (SOD) supplement as a potential therapy for epilepsy, SCN2A expression in the cortex and the correlation between SCN2A and Cu-Zn SOD in SH-SY5Y cells were examined. SCN2A expression and the concentration of Cu-Zn SOD in the cerebral cortexes of patients with primary and secondary temporal lobe epilepsy and normal brain cortex tissues were detected. By transfecting SH-SY5Y cells, the expression of SCN2A and the concentration of Cu-Zn SOD was analyzed and the single-cell patch clamp technique was employed in order to investigate the changes in sodium ion levels following SCN2A knockdown. SCN2A level restoration was also investigated with a Cu-Zn SOD supplement using an expression study and evaluated the changes in sodium ion levels following SCN2A knockdown. SCN2A expression and Cu-Zn SOD concentration decreased in the epileptic cerebral cortex. Following SCN2A knockdown, the concentration of Cu-Zn SOD declined and the si-SCN2A vector group showed a repeated discharge. Furthermore, the Cu-Zn SOD concentration was capable of restoring the expression of SCN2A following SCN2A knockdown in SH-SY5Y cells and the overexpression of Cu-Zn SOD prevented the repeated discharge caused by si-SCN2A. The results indicated that there is a low expression of SCN2A and Cu-Zn SOD in the epileptic cerebral cortex and provided novel insights into potential therapies for temporal lobe epilepsy.
Sigma-Aldrich
Monoclonal Anti-SCN2A1 antibody produced in mouse, clone S69-3, 1 mg/mL, purified immunoglobulin
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Superoxide Dismutase (SOD) antibody produced in mouse, clone SD-G6, ascites fluid
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture