MilliporeSigma
  • Genome-wide identification of miR-200 targets reveals a regulatory network controlling cell invasion.
Genome-wide identification of miR-200 targets reveals a regulatory network controlling cell invasion.
The EMBO journal (2014-07-30)
Cameron P Bracken, Xiaochun Li, Josephine A Wright, David M Lawrence, Katherine A Pillman, Marika Salmanidis, Matthew A Anderson, B Kate Dredge, Philip A Gregory, Anna Tsykin, Corine Neilsen, Daniel W Thomson, Andrew G Bert, Joanne M Leerberg, Alpha S Yap, Kirk B Jensen, Yeesim Khew-Goodall, Gregory J Goodall
ABSTRACT
The microRNAs of the miR-200 family maintain the central characteristics of epithelia and inhibit tumor cell motility and invasiveness. Using the Ago-HITS-CLIP technology for transcriptome-wide identification of direct microRNA targets in living cells, along with extensive validation to verify the reliability of the approach, we have identified hundreds of miR-200a and miR-200b targets, providing insights into general features of miRNA target site selection. Gene ontology analysis revealed a predominant effect of miR-200 targets in widespread coordinate control of actin cytoskeleton dynamics. Functional characterization of the miR-200 targets indicates that they constitute subnetworks that underlie the ability of cancer cells to migrate and invade, including coordinate effects on Rho-ROCK signaling, invadopodia formation, MMP activity, and focal adhesions. Thus, the miR-200 family maintains the central characteristics of the epithelial phenotype by acting on numerous targets at multiple levels, encompassing both cytoskeletal effectors that control actin filament organization and dynamics, and upstream signals that locally regulate the cytoskeleton to maintain cell morphology and prevent cell migration.
Sigma-Aldrich
Monoclonal Anti-ABL2 antibody produced in mouse, clone 5C6, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Vinculin Antibody, clone VIIF9 (7F9), clone VIIF9 (7F9), Chemicon®, from mouse
Sigma-Aldrich
Anti-TKS5 (SH3 #4) Antibody, from rabbit
Sigma-Aldrich
Anti-MPRIP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Anti-MMP-16 Antibody, clone 117-10C6, clone 117-10C6, Chemicon®, from mouse