MilliporeSigma
  • Cloning and characterization of tesk1, a novel spermatogenesis-related gene, in the tongue sole (Cynoglossus semilaevis).
Cloning and characterization of tesk1, a novel spermatogenesis-related gene, in the tongue sole (Cynoglossus semilaevis).
PloS one (2014-10-02)
Liang Meng, Ying Zhu, Ning Zhang, Wanjun Liu, Yang Liu, Changwei Shao, Na Wang, Songlin Chen
ABSTRACT
Testis-specific protein kinase 1 (Tesk1) is a serine/threonine kinase with unique structural features. In the present study, we cloned and characterized the tesk1 gene of tongue sole, Cynoglossus semilaevis. The full-length tesk1 cDNA consists of 1,672 nucleotides, encoding a 331 amino acid polypeptide with a characteristic structure composed of an N-terminal kinase domain and a C-terminal proline-rich domain. The tesk1 genomic sequence contains eight exons and seven introns. Real-time quantitative PCR revealed that tesk1 mRNA is expressed predominantly in the testis, though the level of expression varied throughout development. We used in situ hybridization to show that tesk1 mRNA is expressed in the spermatids of males and pseudo-males, but not in triploid males. Our results suggest that tongue sole Tesk1 may play a role in spermatogenesis.
Product Description
Roche
DIG-Nick Translation Mix, sufficient for 40 labeling reactions, pkg of 160 μL, suitable for hybridization