MilliporeSigma
  • Polymorphism in NFKBIA gene is associated with recurrent acute rejections in liver transplant recipients.
Polymorphism in NFKBIA gene is associated with recurrent acute rejections in liver transplant recipients.
Tissue antigens (2014-08-13)
K Kramer, T Thye, A Treszl, S Peine, M Koch, M Sterneck, B Nashan, H Thude
ABSTRACT
The nuclear factor of kappa light polypeptide gene enhancer B-cells inhibitor-alpha (NFKBIA) gene encodes a member of the nuclear factor-kappa-B inhibitor family. Polymorphisms in this gene might be associated with a susceptibility to acute rejection episodes following liver transplantation, as they may cause an increased activation level of the proinflammatory transcription factor nuclear factor of kappa light polypeptide gene enhancer in B-cells (NFκB). The aim of this study was to evaluate whether the NFKBIA polymorphisms -297 C/T (rs2233409), -826 C/T (rs2233406) and 126 G/A (rs696) affect the incidence of acute liver graft rejection. A total of 199 liver transplant recipients was analyzed, 100 without (NAR) and 99 with early acute rejection (AR). Thirty-two individuals with multiple acute rejections (MAR) were analyzed as a subgroup of AR. Polymerase chain reaction-allele specific restriction enzyme analysis (PCR-ASRA) and allele-specific hybridization with fluorescence resonance energy transfer (FRET) were used for genotyping. We identified the genotype NFKBIA 126 AA (P = 0.002) as well as the haplotype NFKBIA-126A-297T-826T (P = 0.002) as a potential risk factor for the occurrence of recurrent acute rejections. Furthermore, we assessed an association between the 126 A allele and susceptibility to recurrent acute rejections (P = 0.027). Our data suggest that the NFKBIA 126 G/A polymorphism might be potentially helpful to identify liver transplant recipients with an increased susceptibility to develop recurrent acute rejections.