MilliporeSigma
  • Cell-internalization SELEX: method for identifying cell-internalizing RNA aptamers for delivering siRNAs to target cells.
Cell-internalization SELEX: method for identifying cell-internalizing RNA aptamers for delivering siRNAs to target cells.
Methods in molecular biology (Clifton, N.J.) (2014-10-17)
William H Thiel, Kristina W Thiel, Katie S Flenker, Tom Bair, Adam J Dupuy, James O McNamara, Francis J Miller, Paloma H Giangrande
ABSTRACT
After a decade of work to address cellular uptake, the principal obstacle to RNAi-based therapeutics, there is now well-deserved, renewed optimism about RNAi-based drugs. Phase I and II studies have shown safe, strong, and durable-gene knockdown (80-90%, lasting for a month after a single injection) and/or clinical benefit in treating several liver pathologies. Although promising, these studies have also highlighted the need for robust delivery techniques to develop RNAi therapeutics for treating other organ systems and diseases. Conjugation of siRNAs to cell-specific, synthetic RNA ligands (aptamers) is being proposed as a viable solution to this problem. While encouraging, the extended use of RNA aptamers as a delivery tool for siRNAs awaits the identification of RNA aptamer sequences capable of targeting and entering the cytoplasm of many different cell types. We describe a cell-based selection process for the rapid identification and characterization of RNA aptamers suited for delivering siRNA drugs into the cytoplasm of target cells. This process, termed "cell-internalization SELEX (Systematic Evolution of Ligands by Exponential Enrichment)," entails the combination of multiple sophisticated technologies, including cell culture-based SELEX procedures, next-generation sequencing (NGS), and novel bioinformatics tools.
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%