MilliporeSigma
Deuteration of Perylene Enhances Photochemical Upconversion Efficiency.
The journal of physical chemistry letters (2015-08-13)
Andrew Danos, Rowan W MacQueen, Yuen Yap Cheng, Miroslav Dvořák, Tamim A Darwish, Dane R McCamey, Timothy W Schmidt
ABSTRACT
Photochemical upconversion via triplet-triplet annihilation is a promising technology for improving the efficiency of photovoltaic devices. Previous studies have shown that the efficiency of upconversion depends largely on two rate constants intrinsic to the emitting species. Here, we report that one of these rate constants can be altered by deuteration, leading to enhanced upconversion efficiency. For perylene, deuteration decreases the first order decay rate constant by 16 ± 9% at 298 K, which increases the linear upconversion response by 45 ± 21% in the low excitation regime.
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%