MilliporeSigma
  • Polyetherimide-grafted Fe₃O₄@SiO2₂ nanoparticles as theranostic agents for simultaneous VEGF siRNA delivery and magnetic resonance cell imaging.
Polyetherimide-grafted Fe₃O₄@SiO2₂ nanoparticles as theranostic agents for simultaneous VEGF siRNA delivery and magnetic resonance cell imaging.
International journal of nanomedicine (2015-07-15)
Tingting Li, Xue Shen, Yin Chen, Chengchen Zhang, Jie Yan, Hong Yang, Chunhui Wu, Hongjun Zeng, Yiyao Liu
ABSTRACT
Engineering a safe and high-efficiency delivery system for efficient RNA interference is critical for successful gene therapy. In this study, we designed a novel nanocarrier system of polyethyleneimine (PEI)-modified Fe3O4@SiO2, which allows high efficient loading of VEGF small hairpin (sh)RNA to form Fe3O4@SiO2/PEI/VEGF shRNA nanocomposites for VEGF gene silencing as well as magnetic resonance (MR) imaging. The size, morphology, particle stability, magnetic properties, and gene-binding capacity and protection were determined. Low cytotoxicity and hemolyticity against human red blood cells showed the excellent biocompatibility of the multifunctional nanocomposites, and also no significant coagulation was observed. The nanocomposites maintain their superparamagnetic property at room temperature and no appreciable change in magnetism, even after PEI modification. The qualitative and quantitative analysis of cellular internalization into MCF-7 human breast cancer cells by Prussian blue staining and inductively coupled plasma atomic emission spectroscopy analysis, respectively, demonstrated that the Fe3O4@SiO2/PEI/VEGF shRNA nanocomposites could be easily internalized by MCF-7 cells, and they exhibited significant inhibition of VEGF gene expression. Furthermore, the MR cellular images showed that the superparamagnetic iron oxide core of our Fe3O4@SiO2/PEI/VEGF shRNA nanocomposites could also act as a T2-weighted contrast agent for cancer MR imaging. Our data highlight multifunctional Fe3O4@SiO2/PEI/VEGF shRNA nanocomposites as a potential platform for simultaneous gene delivery and MR cell imaging, which are promising as theranostic agents for cancer treatment and diagnosis in the future.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Vegfa
Sigma-Aldrich
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%
Sigma-Aldrich
[3-(2-Aminoethylamino)propyl]trimethoxysilane, technical grade, ≥80%
Sigma-Aldrich
MISSION® esiRNA, targeting human VEGFA