MilliporeSigma
  • The Additive Inflammatory In Vivo and In Vitro Effects of IL-7 and TSLP in Arthritis Underscore the Therapeutic Rationale for Dual Blockade.
The Additive Inflammatory In Vivo and In Vitro Effects of IL-7 and TSLP in Arthritis Underscore the Therapeutic Rationale for Dual Blockade.
PloS one (2015-06-26)
Maarten R Hillen, Sarita A Y Hartgring, Cynthia R Willis, Timothy R D J Radstake, Cornelis E Hack, Floris P J G Lafeber, Joel A G van Roon
ABSTRACT
The cytokines interleukin (IL)-7 and thymic stromal lymphopoietin (TSLP) signal through the IL-7R subunit and play proinflammatory roles in experimental arthritis and rheumatoid arthritis (RA). We evaluated the effect of inhibition of IL-7R- and TSLPR-signalling as well as simultaneous inhibition of IL-7R- and TSLPR-signalling in murine experimental arthritis. In addition, the effects of IL-7 and TSLP in human RA dendritic cell (DC)/T-cell co-cultures were studied. Arthritis was induced with proteoglycan in wildtype mice (WT) and in mice deficient for the TSLP receptor subunit (TSLPR-/-). Both mice genotypes were treated with anti-IL-7R or phosphate buffered saline. Arthritis severity was assessed and local and circulating cytokines were measured. Autologous CD1c-positive DCs and CD4 T-cells were isolated from peripheral blood of RA patients and were co-cultured in the presence of IL-7, TSLP or both and proliferation and cytokine production were assessed. Arthritis severity and immunopathology were decreased in WT mice treated with anti-IL-7R, in TSLPR-/- mice, and the most robustly in TSLPR-/- mice treated with anti-IL-7R. This was associated with strongly decreased levels of IL-17, IL-6 and CD40L. In human DC/T-cell co-cultures, TSLP and IL-7 additively increased T-cell proliferation and production of Th17-associated cytokines, chemokines and tissue destruction factors. TSLP and IL-7 have an additive effect on the production of Th17-cytokines in a human in vitro model, and enhance arthritis in mice linked with enhanced inflammation and immunopathology. As both cytokines signal via the IL-7R, these data urge for IL-7R-targeting to prevent the activity of both cytokines in RA.
Sigma-Aldrich
Dimethyldioctadecylammonium bromide, ≥98% (TLC)
Sigma-Aldrich
TSLP human, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)
Sigma-Aldrich
Dimethyldioctadecylammonium bromide, ≥98.0% (AT)
Sigma-Aldrich
N,N-Dimethyldodecylamine, 97%