MilliporeSigma
  • Hepatic Fibrosis Progression in HIV-Hepatitis C Virus Co-Infection--The Effect of Sex on Risk of Significant Fibrosis Measured by Aspartate-to-Platelet Ratio Index.
Hepatic Fibrosis Progression in HIV-Hepatitis C Virus Co-Infection--The Effect of Sex on Risk of Significant Fibrosis Measured by Aspartate-to-Platelet Ratio Index.
PloS one (2015-06-20)
Kathleen C Rollet-Kurhajec, Erica E M Moodie, Sharon Walmsley, Curtis Cooper, Neora Pick, Marina B Klein
ABSTRACT
In Hepatitis C virus (HCV) mono-infection, male sex is associated with faster liver fibrosis progression but the effects of sex have not been well studied in HIV-HCV co-infected patients. We examined the influence of sex on progression to significant liver fibrosis in HIV-HCV co-infected adults receiving antiretroviral therapy (ART) using the aspartate aminotransferase-to-platelet ratio index (APRI) as a surrogate biomarker of liver fibrosis. We evaluated 308 HIV infected, HCV RNA positive participants of a Canadian multicentre prospective cohort receiving antiretrovirals and without significant liver fibrosis or end-stage liver disease at baseline. We used multivariate discrete-time proportional hazards models to assess the effect of sex on time to significant fibrosis (APRI≥1.5) adjusting for baseline age, alcohol use, cigarette smoking, HCV duration, and APRI and time-updated CD4 count and HIV RNA. Overall, 55 (18%) participants developed an APRI ≥ 1.5 over 544 person-years of at-risk follow-up time; 18 (21%) women (incidence rate (IR)=14.0/100 PY; 7.5-20.4) and 37 (17%) men (IR=8.9/100 PY; 6.0-11.8). Women had more favourable profiles with respect to traditional risk factors for liver disease progression (younger, shorter duration of HCV infection and less alcohol use). Despite this, female sex was associated with a greater than two-fold increased risk of fibrosis progression (adjusted hazard rate (HR) =2.23; 1.22-4.08). HIV-HCV co-infected women receiving antiretroviral therapy were at significantly greater risk of progressing to liver fibrosis as measured by APRI compared with men. Enhanced efforts to engage and treat co-infected women for HCV are needed.
Sigma-Aldrich
Borane dimethyl sulfide complex solution, 2.0 M in THF
Sigma-Aldrich
Borane dimethyl sulfide complex
Sigma-Aldrich
Borane dimethyl sulfide complex solution, 1.0 M in 2-methyltetrahydrofuran