MilliporeSigma
  • Selective chemical modification of DNA with alkoxy- and benzyloxyamines.
Selective chemical modification of DNA with alkoxy- and benzyloxyamines.
Organic & biomolecular chemistry (2015-05-08)
Lorina Gjonaj, Gerard Roelfes
ABSTRACT
A new method for the selective chemical modification of DNA at cytosine nucleobases using alkoxy- and benzyloxyamines is presented. It is shown that in particular benzyloxyamines are effective DNA modifying agents, giving rise to almost exclusive formation of the mono addition products. By using a bifunctional derivative, that is, p-azidobenzyloxyamine hydrochloride, an azide moiety, which is a convenient handle for further functionalization, could be introduced into the DNA. The azido modified DNA was then further reacted in a copper(I)-monophos catalysed 1,3-dipolar cycloaddition. These results illustrate the potential of the presented method for application in site and chemo-selective modification of DNA.
Sigma-Aldrich
Ammonium citrate dibasic, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ammonium citrate dibasic, ACS reagent, 98%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ammonium citrate dibasic, puriss., ≥98%
Sigma-Aldrich
Ammonium citrate dibasic, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis