MilliporeSigma
  • Preparation and in vitro antioxidant activities of 6-amino-6-deoxychitosan and its sulfonated derivatives.
Preparation and in vitro antioxidant activities of 6-amino-6-deoxychitosan and its sulfonated derivatives.
Biopolymers (2015-04-11)
Jianhong Yang, Qinyue Xie, Jianfeng Zhu, Chang Zou, Lingyun Chen, Yumin Du, Dinglong Li
ABSTRACT
The 6-amino-6-deoxychitosan (NC) and their 2, 6-di-N-sulfonated derivatives were prepared via N-phthaloylation, tosylation, azidation, hydrazinolysis, reduction of azide groups and N-sulfonation, and their structures were systematically characterized by FT-IR, 2D HSQC NMR, XRD, gel permeation chromatography (GPC), and elemental analysis. The 6-amino-6-deoxychitosan showed effect in three selected antioxidant essays, including reducing power, superoxide anion radical scavenging ability, and hydroxyl radical scavenging effect. But the factors affecting each activity were different. The reducing power and the superoxide anion radical scavenging ability of NC were strong and closely related to the amino groups in the molecular chains. Both introducing N-sulfonated groups into NC and the concentration reduction of NC and its sulfonated derivatives decreased these activities. For the superoxide anion radical, the molecular charge property was also a significant influence factor. For the hydroxyl radical, NC only showed weak scavenging activity in a special inverse concentration-dependent manner. However, the incorporation of N-sulfonated groups significantly improved the scavenging activity, and the more N-sulfonated groups, the higher the concentrations, the stronger the activity was. The results could be due to the different conformations of NC and its sulfonated derivatives in aqueous solution.
Product Description
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Vinylene carbonate, contains ≤2% BHT as stabilizer, 97%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Sulfur trioxide trimethylamine complex
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt