MilliporeSigma
  • Effects of blockade of endogenous Gi signaling in Tie2-expressing cells on bone formation in a mouse model of heterotopic ossification.
Effects of blockade of endogenous Gi signaling in Tie2-expressing cells on bone formation in a mouse model of heterotopic ossification.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2015-03-17)
Liping Wang, Dylan O' Carroll, Xuhui Liu, Theresa Roth, Hubert Kim, Bernard Halloran, Robert A Nissenson
ABSTRACT
Available evidence indicates that some Tie2-expressing (Tie2(+) ) cells serve as multipotent progenitors that have robust BMP-dependent osteogenic activity and mediate heterotopic ossification (HO). Since signaling through the G protein Gi is required for cell motility, we hypothesized that blockade of endogenous Gi signaling in Tie2(+) cell populations would prevent HO formation. Blockade of Gi signaling in Tie2(+) cells was accomplished in transgenic mice with expression of pertussis toxin (PTX) under the control of the Tie2 promoter (Tie2(+) /PTX(+) ). Bone formation within HOs was evaluated 2 weeks after BMP injection. Expression of PTX in Tie2(+) cells significantly reduced the bone volume (BV) of HOs in male and female mice. Orthotopic bones were assessed at the distal femur and expression of PTX significantly increased trabecular bone fractional volume and bone formation rate in females only. In adult Tie2(+) /GFP(+) mice, GFP(+) cells appeared both inside and at the surfaces of bone tissue within HOs and in orthotopic bones. In summary, blockade of Gi signaling in Tie2(+) cells reduced the accrual of HOs and stimulated osteogenesis in orthotopic bones. Targeting of Gi protein coupled receptors in Tie2(+) cells may be a novel therapeutic strategy in states of abnormal bone formation such as osteoporosis and HO.
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Methyl methacrylate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
Bone Morphogenetic Protein 2 human, Carrier Free, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
BMP-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
BMP-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Tetrazolium Violet
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.