MilliporeSigma
  • Fluorescence emissions of imide compounds and end-capped polyimides enhanced by intramolecular double hydrogen bonds.
Fluorescence emissions of imide compounds and end-capped polyimides enhanced by intramolecular double hydrogen bonds.
Physical chemistry chemical physics : PCCP (2015-11-03)
Kenta Kanosue, Shinji Ando
ABSTRACT
The structure and optical properties of a newly synthesized imide compound (DHNHPI) that forms intramolecular double hydrogen bonds (intra-HBs) were investigated. This compound exhibits intense absorption at 372 nm (ε = 5091 cm(-1) M(-1)) and strong emission at 427 nm (Φ = 0.507) in CHCl3. Under basic conditions, the absorption and fluorescence peaks showed large bathochromic shifts by 70 nm and 95 nm, respectively, compared with the neutral condition due to conversion to anion form. Based on the single-crystal X-ray diffraction analysis, DHNHPI can exert intermolecular π-π interactions between adjacent molecules along the stacking direction, with resultant emission peaks exhibiting longer wavelengths (∼525 nm) in the solid state. Moreover, an end-capped polyimide having DHNHPI moieties at the termini exhibited a strong fluorescence peak at 427 nm by photo-excitation at 332 nm. The large Stokes shift of 6701 cm(-1) clearly indicates the occurrence of an effective Förster-resonance energy transfer from the main chains to the fluorescent end-groups. These facts demonstrate that the incorporation of two -OH groups, which form intra-HBs, endows imide compounds and polyimides with enhanced fluorescence properties as well as high sensitivity towards pH.
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
2,3-Dicyanohydroquinone, 98%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cyclohexylamine, ReagentPlus®, ≥99.9%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
N,N-Dimethylacetamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylacetamide, SAJ special grade
Sigma-Aldrich
Hexachloro-2-propanone, 99%
Sigma-Aldrich
N,N-Dimethylacetamide-d9, 99 atom % D
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexachloro-2-propanone, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)