MilliporeSigma
  • Abnormal dopaminergic modulation of striato-cortical networks underlies levodopa-induced dyskinesias in humans.
Abnormal dopaminergic modulation of striato-cortical networks underlies levodopa-induced dyskinesias in humans.
Brain : a journal of neurology (2015-04-18)
Damian M Herz, Brian N Haagensen, Mark S Christensen, Kristoffer H Madsen, James B Rowe, Annemette Løkkegaard, Hartwig R Siebner
ABSTRACT
Dopaminergic signalling in the striatum contributes to reinforcement of actions and motivational enhancement of motor vigour. Parkinson's disease leads to progressive dopaminergic denervation of the striatum, impairing the function of cortico-basal ganglia networks. While levodopa therapy alleviates basal ganglia dysfunction in Parkinson's disease, it often elicits involuntary movements, referred to as levodopa-induced peak-of-dose dyskinesias. Here, we used a novel pharmacodynamic neuroimaging approach to identify the changes in cortico-basal ganglia connectivity that herald the emergence of levodopa-induced dyskinesias. Twenty-six patients with Parkinson's disease (age range: 51-84 years; 11 females) received a single dose of levodopa and then performed a task in which they had to produce or suppress a movement in response to visual cues. Task-related activity was continuously mapped with functional magnetic resonance imaging. Dynamic causal modelling was applied to assess levodopa-induced modulation of effective connectivity between the pre-supplementary motor area, primary motor cortex and putamen when patients suppressed a motor response. Bayesian model selection revealed that patients who later developed levodopa-induced dyskinesias, but not patients without dyskinesias, showed a linear increase in connectivity between the putamen and primary motor cortex after levodopa intake during movement suppression. Individual dyskinesia severity was predicted by levodopa-induced modulation of striato-cortical feedback connections from putamen to the pre-supplementary motor area (Pcorrected = 0.020) and primary motor cortex (Pcorrected = 0.044), but not feed-forward connections from the cortex to the putamen. Our results identify for the first time, aberrant dopaminergic modulation of striatal-cortical connectivity as a neural signature of levodopa-induced dyskinesias in humans. We argue that excessive striato-cortical connectivity in response to levodopa produces an aberrant reinforcement signal producing an abnormal motor drive that ultimately triggers involuntary movements.
Product Description
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dichloromethane, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)