MilliporeSigma
  • CD39 is a promising therapeutic antibody target for the treatment of soft tissue sarcoma.
CD39 is a promising therapeutic antibody target for the treatment of soft tissue sarcoma.
American journal of translational research (2015-08-19)
Gregory M Hayes, Belinda Cairns, Zoia Levashova, Lawrence Chinn, Myra Perez, Jan-Willem Theunissen, Sindy Liao-Chan, Abel Bermudez, Mark R Flory, Karl J Schweighofer, Edward H van der Horst
ABSTRACT
Soft tissue sarcoma (STS) is a heterogenous tumor arising from the embryonic mesoderm represented by approximately 50 histological subtypes. Effective therapeutic intervention is lacking for recurrent, late stage and metastatic disease. CD39, a cell-surface ectonucleotidase, has previously been shown to be upregulated in hematological malignancies and various epithelial tumors, but not in STS. Here, we show by mass spectrometry and immunohistochemistry that CD39 is highly expressed in primary patient sarcoma samples. Moreover, CD39 nucleotidase activity is enhanced in fibrosarcoma compared with normal control cells. We demonstrate that an inhibitory monoclonal anti-CD39 antibody, abrogates CD39 enzymatic activity significantly and prolongs survival in a lethal metastatic patient-derived sarcoma model. Taken together, the data suggest CD39 is a novel therapeutic target for the treatment of STS.
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium polytungstate, purum p.a., for the preparation of heavy liquid, ≥85% WO3 basis
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Anti-ENTPD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution