MilliporeSigma
  • Purification of Alaskan walleye pollock (Gadus chalcogrammus) and New Zealand hoki (Macruronus novaezelandiae) liver oil using short path distillation.
Purification of Alaskan walleye pollock (Gadus chalcogrammus) and New Zealand hoki (Macruronus novaezelandiae) liver oil using short path distillation.
Nutrients (2014-05-27)
Alex C M Oliveira, Matthew R Miller
ABSTRACT
The beneficial health effects of a diet rich in n-3 long chain polyunsaturated fatty acids (n-3 LC-PUFA) have been extensively researched in recent years. Marine oils are an important dietary source of n-3 LC-PUFA, being especially rich in two of the most important fatty acids of this class, EPA (eicosapentaenoic acid; 20:5n-3) and DHA (docosahexaenoic acid; 22:6n-3). Oils rich in n-3 LC-PUFA are prone to oxidation that leads to loss of product quality. Alaskan pollock (Gadus chalcogrammus Pallas, 1814) and New Zealand's hoki (Macruronus novaezelandiae Hector, 1871) are the highest volume fisheries of their respective countries. Both produce large quantities of fishery byproducts, in particular crude or unrefined n-3 LC-PUFA containing oils. Presently these oils are used as ingredients for animal feed, and only limited quantities are used as human nutritional products. The aim of this research was to investigate the applicability of short path distillation for the purification of pollock and hoki oil to produce purified human-grade fish oil to meet quality specifications. Pollock and hoki oils were subjected to short path distillation and a significant decrease in free fatty acids and lipid oxidation (peroxide and para-anisidine values) products was observed. Purified oils met the Global Organization for EPA and DHA Omega-3 (GOED) standard for edible fish oils.
Sigma-Aldrich
Ascorbic acid 6-palmitate, meets USP testing specifications
Sigma-Aldrich
6-O-Palmitoyl-L-ascorbic acid, BioXtra, ≥99.0% (RT)
Ascorbyl palmitate, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbyl Palmitate, Pharmaceutical Secondary Standard; Certified Reference Material