MilliporeSigma
Protein energy wasting in children with chronic kidney disease.
Pediatric nephrology (Berlin, Germany) (2014-02-08)
Alison G Abraham, Robert H Mak, Mark Mitsnefes, Colin White, Marva Moxey-Mims, Bradley Warady, Susan L Furth
ABSTRACT
In adults with chronic kidney disease (CKD), protein-energy wasting (PEW) is a risk factor for hospitalization and death. However, PEW in children with CKD is not well characterized or defined. Using data from the Chronic Kidney Disease in Children study, we assessed three alternate definitions of PEW using biochemical parameters, body and muscle mass measurements, and reported appetite as described in adults: (1) a minimal PEW definition (≥2 of the four criteria); (2) a standard PEW definition (≥3 of the four criteria); (3) a modified PEW definition (≥3 of the four criteria plus a pediatric-focused criterion of short stature or poor growth). Of the 528 children analyzed in this study (median age 12 years, median glomerular filtration rate 45 mL/min/1.73 m(2), 39% female, 18% African American), 7-20% met the spectrum of definitions for PEW. The unadjusted incidence rates for incident hospitalizations were 1.9-, 2.1-, and 2.2-fold higher for those children diagnosed with PEW using the minimal, standard, and modified definitions, respectively (P = 0.08, 0.09 and 0.03). Following adjustment, only the modified PEW definition, which added short stature or poor growth as a criterion, showed modest significance (P = 0.06). The inclusion of a criterion based on growth may augment the definition of PEW and improve risk discrimination in children with CKD.
Product Description
Iohexol for peak identification, European Pharmacopoeia (EP) Reference Standard
Iohexol, European Pharmacopoeia (EP) Reference Standard
Supelco
Iohexol, analytical standard
Sigma-Aldrich
Histodenz, nonionic density gradient medium