MilliporeSigma
  • Anti-platelet factor 4/heparin antibodies in patients with impaired graft function after liver transplantation.
Anti-platelet factor 4/heparin antibodies in patients with impaired graft function after liver transplantation.
Journal of thrombosis and haemostasis : JTH (2014-03-25)
T Bakchoul, V Assfalg, H Zöllner, M Evert, A Novotny, E Matevossian, H Friess, D Hartmann, G Hron, K Althaus, A Greinacher, N Hüser
ABSTRACT
Heparin, the standard perioperative anticoagulant for the prevention of graft vessel thrombosis in patients undergoing liver transplantation (LT), binds to the chemokine platelet factor 4 (PF4). Antibodies that are formed against the resulting PF4/heparin complexes can induce heparin-induced thrombocytopenia. LT is a clinical situation that allows the study of T-cell dependency of immune responses because T-cell function is largely suppressed pharmacologically in these patients to prevent graft rejection. To investigate the immune response against PF4/heparin complexes in patients undergoing LT. In this prospective cohort study, 38 consecutive patients undergoing LT were systematically screened for anti-PF4/heparin antibodies (enzyme immunoassay and heparin-induced platelet aggregation assay), platelet count, liver function, and engraftment. At baseline, 5 (13%) of 38 patients tested positive for anti-PF4/heparin IgG (non-platelet-activating) antibodies. By day 20, an additional 5 (15%) of 33 patients seroconverted for immunoglobulin G (two platelet-activating) antibodies. No patient developed clinical heparin-induced thrombocytopenia. Two of six patients with graft function failure had anti-PF4/heparin IgG antibodies at the time of graft function failure. Graft liver biopsy samples from these patients showed thrombotic occlusions of the microcirculation. Anti-PF4/heparin IgG antibodies are generated despite strong pharmacologic suppression of T cells, indicating that T cells likely have a limited role in the immune response to PF4/heparin complexes in humans.
Product Description
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
Prednisolone, European Pharmacopoeia (EP) Reference Standard
Supelco
Prednisolone, VETRANAL®, analytical standard
Sigma-Aldrich
Prednisolone, ≥99%
USP
Prednisolone, United States Pharmacopeia (USP) Reference Standard
Prednisolone, British Pharmacopoeia (BP) Assay Standard
Prednisolone for peak identification, European Pharmacopoeia (EP) Reference Standard