MilliporeSigma
  • Direct cytosolic siRNA delivery by reconstituted high density lipoprotein for target-specific therapy of tumor angiogenesis.
Direct cytosolic siRNA delivery by reconstituted high density lipoprotein for target-specific therapy of tumor angiogenesis.
Biomaterials (2014-05-31)
Yang Ding, Yazhe Wang, Jianping Zhou, Xiaochen Gu, Wei Wang, Congyan Liu, Xiuli Bao, Cheng Wang, Yuanru Li, Qiang Zhang
ABSTRACT
We described here the mechanisms by which small interfering RNA (siRNA) molecules incorporated in reconstituted high density lipoprotein (rHDL) were efficiently transferred into the cytoplasm of cells to perform target-specific therapy of tumor angiogenesis. Using fluorescent-tagged apolipoprotein A-I (apoA-I) and cholesterol-conjugated siRNA (Chol-siRNA), it was confirmed with FACS and confocal microscopic measurements that Chol-siRNA-loaded rHDL nanoparticles (rHDL/Chol-siRNA complexes) were successfully established and apoA-I certainly was attached to the surface of Chol-siRNA-loaded lipoplexes (Lipos/Chol-siRNA complexes). Stably assembled rHDL/Chol-siRNA complexes demonstrated proper nanosize, quasi-spherical shape and improved nuclease protection over naked Chol-siRNA. It was also interesting to note that rHDL provided a highly effective approach to transfer Chol-siRNA across the membrane directly into the cytoplasm via the scavenger receptor BI (SR-BI)-mediated non-endocytotic mechanism, thereby bypassing endo-lysosomal trapping. We also showed clear evidence that the in vitro implementation of rHDL for Chol-siRNA-VEGF (Chol-siRNA targeting vascular endothelial growth factor gene) delivery markedly promoted RNA interference (RNAi)-mediated degradation of VEGF mRNA, resulting in down-regulation of secreted VEGF protein. In vivo fluorescence imaging indicated that near-infrared (NIR) dye Cy5 labeled Chol-siRNA-loaded rHDL nanoparticles (rHDL/Cy5-Chol-siRNA complexes) displayed long circulation time, SR-BI positive tumor-selective targeting, and efficient cytosolic delivery capabilities. Furthermore, intravenous administration of Chol-siRNA-VEGF-loaded rHDL nanoparticles (rHDL/Chol-siRNA-VEGF complexes) significantly enhanced anti-tumor efficacy against breast cancer, decreased VEGF expression level, and inhibited formation of intratumoral microvessels at the tumor tissue. It was concluded that rHDL possessed therapeutic potential and versatility in mediating Chol-siRNA-VEGF direct cytosolic delivery for target-specific anti-angiogenic therapy in breast cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human VEGFA
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Vegfa