MilliporeSigma
  • Controlled-release triple anti-inflammatory therapy based on novel gastroretentive sponges: characterization and magnetic resonance imaging in healthy volunteers.
Controlled-release triple anti-inflammatory therapy based on novel gastroretentive sponges: characterization and magnetic resonance imaging in healthy volunteers.
International journal of pharmaceutics (2014-06-15)
Mina Ibrahim Tadros, Rania Hassan Fahmy
ABSTRACT
The current work aimed to develop novel composite sponges of chitosan (CH)-chondroitin sulfate (CS) as a low-density gastroretentive delivery system for lornoxicam (LOR). This triple anti-inflammatory therapy-loaded matrices are expected to expand and float upon contact with gastric fluids for prolonged times. CH and CS solutions (3%, w/w) were prepared, mixed in different ratios, lyophilized, coated with magnesium stearate and compressed. The CH:CS interpolymer complex (IPC) was evaluated via FT-IR, DSC, and XRD. The compressed-sponges were evaluated for appearance, structure, porosity, pore diameter, density, wetting-time, floating characteristics, adhesion-retention, and LOR-release. The gastroretentivity of the best achieved magnetite-loaded sponges was monitored in healthy volunteers via MRI. The interaction between CH (protonated amino groups) and CS (anionic carboxylate/sulfate groups) proved IPC formation. DSC and XRD studies confirmed loss of LOR crystallinity. The sponges possessed interconnecting porous-network structures. The porosity, mean pore diameter, and bulk density of CH:CS (10:3) IPC sponges were 11.779%, 25.4 nm, and 0.670 g/mL, respectively. They showed complete wetting within seconds, gradual size-expansion within minutes and prolonged adhesion for hours. Controlled LOR-release profiles were tailored over 12h to satisfy individual patient needs. Monitoring of sponges via MRI proved their gastroretentivity for at least 5h.
Sigma-Aldrich
D-Lactose monohydrate, SAJ special grade
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
D-Lactose monohydrate, ≥99% (HPLC), BioUltra
Sigma-Aldrich
D-Lactose monohydrate, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Iron(III) oxide, hydrated, catalyst grade, 30-50 mesh
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Lactose (Monohydrate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(III) oxide, SAJ first grade, ≥98.0%