MilliporeSigma
  • Tissue-smashing based ultra-rapid extraction of chemical constituents in herbal medicines.
Tissue-smashing based ultra-rapid extraction of chemical constituents in herbal medicines.
Journal of pharmaceutical and biomedical analysis (2014-04-02)
Yong Fan, Chen-Pu Yan, Cheng Chen, Kwok-Fai So, Ping Li, Lian-Wen Qi
ABSTRACT
Sample extraction is the first challenge in analysis of herbal medicines (HMs). Numerous methods have been developed to improve extraction efficiency, use less solvent and short time. In this work, a tissue-smashing based ultra-rapid extraction (TSURE) method has been proposed through the designed particle crushing, drastic stir, and dynamic molecular permeation at normal temperature. Factors in TSURE like extraction time, volts, and solvents were optimized for extraction efficiency of salvianolic acid B, cryptotanshinone, and tanshinone IIA from Salvia miltiorrhiza. The TSURE method was validated in terms of repeatability (RSD<2.2%) and extraction recoveries (93-106% with RSD<5.0%). TSURE showed a comparable extraction efficiency to conventional heat reflux extraction (HRE) and better than ultrasonic assisted extraction (UAE). The extraction time was about 2.0-3.0 min for TSURE, 60 times faster than the performance of HRE and 20 times faster than UAE. Microscopic analysis showed that the Krummbein diameter of plant particles after extraction were about 600-1200 μm for HRE and UAE, and decreased to 50-80 μm for TSURE. Subsequently, the developed TSURE was applied to high-throughput extraction of 19 S. miltiorrhiza samples collected in different regions of China. Besides, application of TSURE to other herbal medicines was also investigated, including Panax quinquefolius and Lonicera japonica. TSURE method provided an ultra-rapid and promising alternation for extraction of ingredients in herbal medicines, and can be extended to pharmaceutics, foods and cosmetics.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Chlorogenic acid, primary reference standard
Supelco
Salvianolic acid B, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Salvianolic acid B, ≥94% (HPLC)
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Salvianolic acid B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard