MilliporeSigma
Measurement of plasma protein and lipoprotein binding of pyrethroids.
Journal of pharmacological and toxicological methods (2014-06-15)
Pankaj K Sethi, S Muralidhara, James V Bruckner, Catherine A White
ABSTRACT
A simple, reliable procedure was developed to measure binding of pyrethroid insecticides to total proteins and lipoproteins of rat and human plasma. The extent of binding of (14)C-labeled deltamethrin (DLM), cis-permethrin (CIS) and trans-permethrin (TRANS) was quantified by a 3-step organic solvent extraction technique. Rat and human plasma samples, containing NaF to inhibit esterases, were spiked with a range of concentrations of each radiolabeled pyrethroid. Protein binding reached equilibrium within ~1h of incubation at 37°C. The samples were extracted in turn with: isooctane to collect the unbound fraction; 2-octanol to extract the lipoprotein-bound fraction; and acetonitrile to obtain the protein-bound fraction. Absolute recoveries of DLM, CIS and TRANS ranged from 86 to 95%. Adherence of these very lipophilic chemicals to glass and plastic was minimized by using silanized glass vials and LoBind® plastic pipettes. The method's ability to distinguish lipoprotein from protein binding was confirmed by experiments with diazepam and cyclosporine, drugs that bind selectively to albumin and lipoproteins, respectively. This procedure was effectively utilized for studies of the species-dependence of plasma protein and lipoprotein binding of three pyrethroids for inclusion in physiologically-based pharmacokinetic models of pyrethroids for use in health risk assessments of the insecticides in children and adults.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
2-Octanol, ≥97%, FG
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Octanol, 97%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
2,2,4-Trimethylpentane, JIS special grade
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
2,2,4-Trimethylpentane, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
2,2,4-Trimethylpentane, SAJ first grade, ≥98.0%
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Sigma-Aldrich
(±)-2-Octanol, ReagentPlus®, ≥99.5% (GC)
Supelco
2,2,4-Trimethylpentane, analytical standard
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%