MilliporeSigma
  • Gene-based association identifies SPATA13-AS1 as a pharmacogenomic predictor of inhaled short-acting beta-agonist response in multiple population groups.
Gene-based association identifies SPATA13-AS1 as a pharmacogenomic predictor of inhaled short-acting beta-agonist response in multiple population groups.
The pharmacogenomics journal (2014-01-15)
B Padhukasahasram, J J Yang, A M Levin, M Yang, E G Burchard, R Kumar, P-Y Kwok, M A Seibold, D E Lanfear, L K Williams
ABSTRACT
Inhaled short-acting beta-agonist (SABA) medication is commonly used in asthma patients to rapidly reverse airway obstruction and improve acute symptoms. We performed a genome-wide association study of SABA medication response using gene-based association tests. A linear mixed model approach was first used for single-nucleotide polymorphism associations, and the results were later combined using GATES to generate gene-based associations. Our results identified SPATA13-AS1 as being significantly associated with SABA bronchodilator response in 328 healthy African Americans. In replication, this gene was associated with SABA response among the two separate groups of African Americans with asthma (n=1073, P=0.011 and n=1968, P=0.014), 149 healthy African Americans (P=0.003) and 556 European Americans with asthma (P=0.041). SPATA13-AS1 was also associated with longitudinal SABA medication usage in the two separate groups of African Americans with asthma (n=658, P=0.047 and n=1968, P=0.025). Future studies are needed to delineate the precise mechanism by which SPATA13-AS1 may influence SABA response.
Product Description
Supelco
Albuterol sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Salbutamol sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Salbutamol hemisulfate salt, ≥98%