MilliporeSigma
  • Determination of pergolide in horse plasma by UPLC-MS/MS for pharmacokinetic applications.
Determination of pergolide in horse plasma by UPLC-MS/MS for pharmacokinetic applications.
Journal of pharmaceutical and biomedical analysis (2014-02-20)
Glenn A Jacobson, Adam Pirie, Scott Edwards, Kristopher J Hughes, David I Rendle, Noel W Davies
ABSTRACT
Pergolide, an ergot-derived dopamine D2 receptor agonist, is used extensively as an orally administered treatment for pituitary pars intermedia dysfunction (PPID) in horses. One of the barriers associated with pergolide determinations in plasma for pharmacokinetic applications has been the technically demanding requirement for sensitivity. The objective of our work was to develop a simple assay for the determination of pergolide in plasma and demonstrate its potential application in the study of pergolide pharmacokinetics (PK) in horses. A UPLC-MS/MS assay was developed with a simple sample preparation involving methanol protein precipitation and injection of supernatant. The assay was applied to samples from a horse dosed with 10mg pergolide (as the mesylate salt) by nasogastric intubation. Plasma samples were collected over a 48h period. The assay demonstrated performance sufficient to enable application to low level PK studies. Within-batch precision and accuracy were within acceptance criteria; precision was less than 10% RSD (n=5) and accuracy was -7.3% at 0.014ng/mL, the lower limit of quantification was 0.006ng/mL and the method detection limit was 0.002ng/mL. In the treated horse, Cmax was 0.40ng/mL and the assay easily allowed determination of plasma levels in the elimination phase to 48h. In conclusion, this assay using UPLC-MS/MS and methanol protein precipitation easily meets the challenging demands of pergolide analyses in plasma.
Product Description
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)
Pergolide mesilate, European Pharmacopoeia (EP) Reference Standard