MilliporeSigma
  • Application of an optosensing chip based on molecularly imprinted polymer coated quantum dots for the highly selective and sensitive determination of sesamol in sesame oils.
Application of an optosensing chip based on molecularly imprinted polymer coated quantum dots for the highly selective and sensitive determination of sesamol in sesame oils.
Journal of agricultural and food chemistry (2015-02-24)
Huilin Liu, Dan Wu, Yingli Liu, Huijuan Zhang, Tiezheng Ma, Ainasi Aidaerhan, Jing Wang, Baoguo Sun
ABSTRACT
A novel optosensing chip was constructed by anchoring the fluorescence sensing material layer based on molecularly imprinted polymer (MIP) coated CdSe/ZnS quantum dots (QDs) on a chip for highly selective and sensitive optosensing of sesamol in sesame oil. Many factors that affected the performance of the optosensing chip based on MIP-coated QDs are discussed. Under optimized conditions, the relative fluorescence intensity of the optosensing chip decreased linearly (r(2) > 0.99) with increasing sesamol concentration in the range from 2.4 × 10(-6) to 1.2 × 10(-3) mol L(-1) with a detection limit of 7.2 × 10(-8) mol L(-1). The relative standard deviation for five replicate detections of 4.8 × 10(-4) mol L(-1) sesamol was 2.2%. Recoveries of 94.8-102.3% were achieved by direct detection when the optosensing chip was used for the selective detection of sesamol in sesame oil. Practically, the optosensing approach showed high sensitivity, good selectivity, and excellent reproducibility for the detection of sesamol in real oil samples.
Product Description
Supelco
3-tert-Butyl-4-hydroxyanisole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
tert-Butylhydroquinone, 97%
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Sigma-Aldrich
Methacrylic acid, SAJ first grade, ≥98.0%
Sigma-Aldrich
Piperonal, ≥99%, FCC, FG
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Piperonal, 99%
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)