MilliporeSigma
  • Physico-chemical characterization and biological evaluation of two fibroin materials.
Physico-chemical characterization and biological evaluation of two fibroin materials.
Journal of tissue engineering and regenerative medicine (2012-09-14)
Antonella Motta, Paola Segnana, Lucia Verin, Silvia La Monica, Claudia Fumarola, Giovanna Bucci, Francesca Gussago, Anna Maria Cantoni, Luca Ampollini, Claudio Migliaresi
ABSTRACT
Silk fibroin fibres from two different sources, Bombyx mori pure-breed silkworms and polyhybrid cross-bred silkworm cocoons, were treated with formic acid under planar stirring conditions to prepare non-woven nets. The treatment partially dissolved the fibres, which bound together and formed a non-woven micrometric net with fibres coated by a thin layer of low molecular weight fibroin matrix. The starting fibres, net materials and fibroin coating layer were characterized in terms of amino acid composition, molecular weight and calorimetric properties. In vitro cell culture tests with rat fibroblasts were performed to investigate cell proliferation, morphology and spreading. Moreover, host-rat fibroblasts were preseeded on the afore-mentioned nets and implanted in the thorax of rats for histological analysis. In spite of the chemical differences between the two starting fibroins, the response of the said materials in vitro and in vivo were very similar. These results suggest that the outcome is likely correlated with the modification of the processing technique; that during the formation of the net, a thin gel layer of similar amino acid composition was formed on the fibroin fibres.
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%