MilliporeSigma
  • Hierarchically porous three-dimensional electrodes of CoMoO₄ and ZnCo₂O₄ and their high anode performance for lithium ion batteries.
Hierarchically porous three-dimensional electrodes of CoMoO₄ and ZnCo₂O₄ and their high anode performance for lithium ion batteries.
Nanoscale (2014-08-15)
Hong Yu, Cao Guan, Xianhong Rui, Bo Ouyang, Boluo Yadian, Yizhong Huang, Hua Zhang, Harry E Hoster, Hong Jin Fan, Qingyu Yan
ABSTRACT
Ternary metal oxides have been receiving wide attention in electrochemical energy storage due to their rich redox reactions and tuneable conductivity. We present a simple solution-based method to prepare a 3D interconnected porous network of ternary metal oxide (CoMoO₄ and ZnCo₂O₄) nanostructures on macroporous nickel foam. The open-structured networks with different degrees of porosity endow them with high surface areas of electro-active sites. The Li ion storage properties of both anodes are investigated. High rate capability and long term cycling stability are achieved for both systems.
Sigma-Aldrich
Hexamethylenetetramine, JIS special grade, ≥99.0%
Sigma-Aldrich
Hexamethylenetetramine, meets analytical specification of Ph. Eur., BP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hexamethylenetetramine, SAJ first grade, ≥98.5%
Sigma-Aldrich
Hexamethylenetetramine, ReagentPlus®, 99%
Sigma-Aldrich
Hexamethylenetetramine, ACS reagent, ≥99.0%
Sigma-Aldrich
Hexamethylenetetramine, puriss. p.a., reag. Ph. Eur., ≥99.5% (calc. to the dried substance)