MilliporeSigma
  • Harnessing medically relevant metals onto water-soluble subphthalocyanines: towards bimodal imaging and theranostics.
Harnessing medically relevant metals onto water-soluble subphthalocyanines: towards bimodal imaging and theranostics.
Dalton transactions (Cambridge, England : 2003) (2015-01-13)
Yann Bernhard, Pascale Winckler, Jean-Marie Perrier-Cornet, Richard A Decréau
ABSTRACT
Subphthalocyanine (SubPc), a putative fluorophore for optical imaging (OI), was conjugated to chelating ligands (DOTA, DTPA) affording water-soluble conjugates complexed with (non-radioactive) metals relevant to the following medical imaging techniques/therapies: MRI (Gd), PET (Cu, Ga), SPECT (In, Ga, Lu), RIT (Cu, Lu, Y), and NCT (Gd). Magneto-optical properties of ditopic gadolinium species (and optical properties of other metal containing species) were examined (brightness (ε × ΦF) and relaxivity R1) and fluorescence confocal/biphoton microscopy studies were conducted.
Product Description
Dithranol, European Pharmacopoeia (EP) Reference Standard
Supelco
2,5-Dihydroxybenzoic acid, analytical standard
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Sigma-Aldrich
Dithranol, ≥90% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure