MilliporeSigma
  • Targeting galectin-1-induced angiogenesis mitigates the severity of endometriosis.
Targeting galectin-1-induced angiogenesis mitigates the severity of endometriosis.
The Journal of pathology (2014-07-01)
Juan I Bastón, Rosa I Barañao, Analía G Ricci, Mariela A Bilotas, Carla N Olivares, José J Singla, Alejandro M Gonzalez, Juan C Stupirski, Diego O Croci, Gabriel A Rabinovich, Gabriela F Meresman
ABSTRACT
Endometriosis is characterized by the presence of endometrial tissue outside the uterus that causes severe pelvic pain and infertility in women of reproductive age. Although not completely understood, the pathophysiology of the disease involves chronic dysregulation of inflammatory and vascular signalling. In the quest for novel therapeutic targets, we investigated the involvement of galectin-1 (Gal-1), an endogenous glycan-binding protein endowed with both immunosuppressive and pro-angiogenic activities, in the pathophysiology of endometriotic lesions. Here we show that Gal-1 is selectively expressed in stromal and endothelial cells of human endometriotic lesions. Using an experimental endometriosis model induced in wild-type and Gal-1-deficient (Lgals1(-/-) ) mice, we showed that this lectin orchestrates the formation of vascular networks in endometriotic lesions in vivo, facilitating their ectopic growth independently of vascular endothelial growth factor (VEGF) and the keratinocyte-derived CXC-motif (CXC-KC) chemokine. Targeting Gal-1 using a specific neutralizing mAb reduced the size and vascularized area of endometriotic lesions within the peritoneal compartment. These results underline the essential role of Gal-1 during endometriosis and validate this lectin as a possible target for the treatment of disease.
Product Description
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Rat CINC-1 ELISA Kit, for serum, plasma and cell culture supernatant
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Human Procalcitonin ELISA Kit
Sigma-Aldrich
Human GRO-α / CXCL1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Mouse KC / CXCL1 ELISA Kit, for serum, plasma and cell culture supernatant
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
o-Xylene, SAJ special grade, ≥98.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
o-Xylene, anhydrous, 97%