MilliporeSigma
  • Novel analogs of antitumor agent calixarene 0118: Synthesis, cytotoxicity, click labeling with 2-[(18)F]fluoroethylazide, and in vivo evaluation.
Novel analogs of antitumor agent calixarene 0118: Synthesis, cytotoxicity, click labeling with 2-[(18)F]fluoroethylazide, and in vivo evaluation.
European journal of medicinal chemistry (2014-12-03)
Tilman Läppchen, Ruud P M Dings, Raffaella Rossin, Justine F Simon, Ton J Visser, Martine Bakker, Priya Walhe, Tiemen van Mourik, Katia Donato, Judy R van Beijnum, Arjan W Griffioen, Johan Lub, Marc S Robillard, Kevin H Mayo, Holger Grüll
ABSTRACT
Calixarene 0118 is a potent anti-angiogenic agent that effectively inhibited tumor growth in preclinical studies, and is currently being evaluated in a phase I clinical trial. We have designed two close mimetics of calixarene 0118 containing a terminal alkynyl-functional group, and developed an optimized semi-automated procedure for radiolabeling with 2-[(18)F]fluoroethylazide using click chemistry. Following semi-preparative HPLC purification and formulation, the lower-rim modified analog [(18)F]6 and the equatorially labeled [(18)F]13 were obtained in >97% radiochemical purity and overall decay-corrected isolated radiochemical yields of 18.7 ± 2.7% (n = 4) and 10.2 ± 5.0% (n = 4), respectively, in a total synthesis time of about 2 h. Preliminary in vivo studies in nude mice bearing human tumor xenografts revealed highest accumulation of both tracers in the liver, followed by spleen, kidney, lung and bone, with no substantial uptake in the tumor. Still, these first-in-class radiotracers are a valuable tool for pharmacokinetic profiling and improvement of calixarene-based anti-angiogenic therapeutics in the future, as similar radiolabeling strategies may be applied to other compounds in the calixarene series. The cold reference compounds of the radiotracers were characterized in terms of cytotoxicity and anti-proliferative effects on HUVEC cells and on MA148 human ovarian carcinoma cells, along with the respective precursors, a small series of 0118 analogs modified with short-chain linear alkyl substituents, and a PEG3-spaced calixarene dimer. While all of the new analogs proved at least equipotent to parent 0118, some of them inhibited HUVEC and MA148 cell growth almost 4- and 10-fold more effectively, rendering these analogs promising candidates for further evaluation in anti-angiogenic cancer therapy.
Sigma-Aldrich
1,11-Diazido-3,6,9-trioxaundecane
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Polyoxyethylene bis(azide) solution, average Mn 450, 0.1 g/mL in THF
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, HPLC grade, ≥99.93%