MilliporeSigma
  • Diosmin pretreatment improves cardiac function and suppresses oxidative stress in rat heart after ischemia/reperfusion.
Diosmin pretreatment improves cardiac function and suppresses oxidative stress in rat heart after ischemia/reperfusion.
European journal of pharmacology (2014-04-29)
Oomaidurai Senthamizhselvan, Jeganathan Manivannan, Thangarasu Silambarasan, Boobalan Raja
ABSTRACT
Reperfusion of ischemic tissue leads to the generation of oxygen derived free radicals which plays an important role in cellular damage. Objective of the current study is to evaluate the cardio-protective and antioxidant effect of diosmin on ischemia-reperfusion related cardiac dysfunction, oxidative stress and apoptosis. Diosmin (50 and 100 mg/kg body weight (bw)) was given every day to the rats orally throughout the experimental period. Ischemia/reperfusion protocol was carried out ex vivo using langendorff perfusion method and the cardiac functional recovery was assessed in terms of percentage rate pressure product. Coronary effluents of LDH and CK-MB activities, antioxidant enzyme activities, lipid peroxidation products, activity of TCA cycle enzymes were evaluated. Moreover, in vitro superoxide anion and hydroxyl radical scavenging potential of diosmin was also quantified. Finally, quantitative real-time PCR was used for assessing Bcl-2 mRNA expression in heart. Cardiac functional recovery was impaired after reperfusion compared with continuously perfused heart. It was significantly prevented by diosmin treatment. Impaired antioxidant enzyme activities and elevated lipid peroxidation products level were also significantly suppressed. The activity of TCA cycle enzymes was protected against reperfusion stress. Down regulated Bcl-2 was also significantly increased. This study concluded that diosmin pretreatment prevents all the impaired patterns including cardiac function, oxidative stress and apoptosis associated with reperfusion in control heart by its antioxidant role.
Product Description
Diosmin for system suitability, European Pharmacopoeia (EP) Reference Standard
Diosmin, European Pharmacopoeia (EP) Reference Standard
Supelco
Diosmin, analytical standard