MilliporeSigma
  • Nanofiber-coated drug eluting stent for the stabilization of mast cells.
Nanofiber-coated drug eluting stent for the stabilization of mast cells.
Pharmaceutical research (2014-03-26)
Byeongtaek Oh, Chi H Lee
ABSTRACT
The nanofiber-hydrogel blend containing nitric oxide (NO) donors and reactive oxygen species (ROS) scavengers (Edaravone: EDV) was explored as an advanced strategy for stabilization of Mast cells (MCs) to achieve efficient immune-suppressive effects. Three types of nanofiber hydrogel composites (Bare-Nanofibers (BNF), Nanofiber-Hydrogels (NF-Gel) and Cross-linked Nanofiber-Hydrogels (NF-Gel-X)), were evaluated. The degranulation rates of MCs were determined by measurement of the extracellular levels of hydrogen peroxide and the released amounts of β-hexosaminidase from the activated-MCs (a-MCs). In addition, the effects of EDV on the selective scavenging of the oxygen radicals and prevention of peroxynitrite formation were evaluated. The roles of a-MCs in re-endothelialization and viability of coronary artery endothelial cells (hPCAECs) were defined using alamar blue and LDH assay, respectively. Each polymer matrix has unique morphological characteristics. The effects of EDV (~1.0 mM) on the production of NO were greatly influenced by the presence of superoxide or hydroxyl radicals. NF-G-X containing a mixture of EDV and S-Nitroglutathione (GSNO) produced the highest level of NO under the oxidative stress conditions. GSNO alone or a mixture of GSNO and EDV significantly lowered the degranulation rate of a-MCs (GSNO only: 55.8 ± 5.4%; GSNO with EDV: 50.6 ± 0.6%), indicating that NO plays an integral role in degranulation of a-MCs. There were no significant biochemical evidences of cytotoxic effects of GSNO and EDV on the hPCAECs. Nanofibers containing a mixture of nitric oxide donors and ROS scavengers could be used as a promising strategy to stabilize MCs from the ROS-mediated immune responses.
Product Description
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Phenazone impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%