MilliporeSigma
  • Differential role of AMPA receptors in mouse tests of antidepressant and anxiolytic action.
Differential role of AMPA receptors in mouse tests of antidepressant and anxiolytic action.
Brain research (2015-01-13)
Jesper T Andreasen, Ciaran M Fitzpatrick, Maria Larsen, Lars Skovgaard, Simon D Nielsen, Rasmus P Clausen, Karin Troelsen, Darryl S Pickering
ABSTRACT
Depression and anxiety often co-occur, and conventional monoamine-facilitating antidepressants show efficacy against symptoms in both disorders. Rodent studies indicate that antidepressant effects of monoamine-based antidepressants involve increased α-amino-3-hydroxy-5- methyl-4-isoxazolepropionic acid glutamate receptor (AMPAR) neurotransmission, and positive allosteric modulators (PAMs) at AMPARs produced antidepressant-like effects in rodents. While this suggests that increased AMPAR-mediated neurotransmission is beneficial in depression management, preclinical studies addressing AMPARs in relation to anxiety have given ambiguous results with both anxiolytic-like and anxiogenic-like effects observed after AMPAR blockade. This study systematically compared the effects of the AMPAR potentiator LY451646 and the AMPAR antagonist GYKI-53655 on depression-related behaviour using the mouse forced swim (FST) and tail suspension tests (TST), and anxiety-related behaviour using the elevated zero maze (EZM), marble burying (MB) and novelty-induced hypophagia (NIH) tests. The serotonin-selective antidepressant citalopram was included for comparison. Due to the importance of AMPARs in learning and memory we also tested if GYKI-53655 disrupted performance in the V-maze test for attention-dependent behaviour, and the social transmission of food preference (STFP) test of long-term memory. LY451646 (3 mg/kg) showed an antidepressant-like profile in the FST and TST, and GYKI-53655 (≥ 5 mg/kg) had a depressogenic-like effect in the TST but no effect in the FST. Conversely, GYKI-53655 produced marked anxiolytic-like effects in the EZM (≥ 2.5 mg/kg), MBT (≥ 2.5 mg/kg), and NIH tests (≥ 5 mg/kg), while LY451646 (≥ 3 mg/kg) increased anxiety-like behaviour in the EZM. Citalopram showed an antidepressant-like effect in the FST (≥ 10 mg/kg), but not TST, an anxiolytic-like effect in the EZM (≥ 3 mg/kg) and MB test (≥ 2.5 mg/kg), and an anxiogenic-like effect in the NIH test (≥ 30 mg/kg). GYKI-53655 did not affect cognitive performance in the V-maze or STFP tests. Collectively, these findings suggest a differential role of AMPARs in depression and anxiety, with AMPAR activation promoting antidepressant responses and AMPAR inhibition promoting anxiolytic responses. The potential of AMPARs as a novel target in depression and anxiety pharmacotherapy is discussed.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Citalopram for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
D-(+)-Glucose monohydrate, meets analytical specification of Ph. Eur., BP, Ph Franç., 97.5-102.0% anhydrous basis(HPLC)
Sigma-Aldrich
D-(+)-Glucose monohydrate, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Millipore
D-(+)-Glucose monohydrate, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Citalopram hydrobromide, ≥98% (HPLC)
Sigma-Aldrich
Dextrose monohydrate, 97.5-102.0% anhydrous basis, meets USP testing specifications