MilliporeSigma
  • A novel TLR-9 agonist C792 inhibits plasmacytoid dendritic cell-induced myeloma cell growth and enhance cytotoxicity of bortezomib.
A novel TLR-9 agonist C792 inhibits plasmacytoid dendritic cell-induced myeloma cell growth and enhance cytotoxicity of bortezomib.
Leukemia (2014-01-31)
A Ray, Z Tian, D S Das, R L Coffman, P Richardson, D Chauhan, K C Anderson
ABSTRACT
Our prior study in multiple myeloma (MM) patients showed increased numbers of plasmacytoid dendritic cells (pDCs) in the bone marrow (BM), which both contribute to immune dysfunction as well as promote tumor cell growth, survival and drug resistance. Here we show that a novel Toll-like receptor (TLR-9) agonist C792 restores the ability of MM patient-pDCs to stimulate T-cell proliferation. Coculture of pDCs with MM cells induces MM cell growth; and importantly, C792 inhibits pDC-induced MM cell growth and triggers apoptosis. In contrast, treatment of either MM cells or pDCs alone with C792 does not affect the viability of either cell type. In agreement with our in vitro data, C792 inhibits pDC-induced MM cell growth in vivo in a murine xenograft model of human MM. Mechanistic studies show that C792 triggers maturation of pDCs, enhances interferon-α and interferon-λ secretion and activates TLR-9/MyD88 signaling axis. Finally, C792 enhances the anti-MM activity of bortezomib, lenalidomide, SAHA or melphalan. Collectively, our preclinical studies provide the basis for clinical trials of C792, either alone or in combination, to both improve immune function and overcome drug resistance in MM.
Sigma-Aldrich
Human TRAIL R2 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
NGF-β human, from human, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Mouse Tnfsf10 / Tumor Necrosis Factor Ligand Superfamily Member 10 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Human TRAIL R4 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Human TRAIL R3 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Melphalan, powder
Sigma-Aldrich
SAHA, ≥98% (HPLC)