MilliporeSigma
  • Urinary elimination of coproporphyrins is dependent on ABCC2 polymorphisms and represents a potential biomarker of MRP2 activity in humans.
Urinary elimination of coproporphyrins is dependent on ABCC2 polymorphisms and represents a potential biomarker of MRP2 activity in humans.
Journal of biomedicine & biotechnology (2011-05-05)
Isabelle Benz-de Bretagne, Renaud Respaud, Patrick Vourc'h, Jean-Michel Halimi, Agnès Caille, Jean-Sébastien Hulot, Christian R Andres, Chantal Le Guellec
ABSTRACT
MRP2 encoded by ABCC2 gene is involved in the secretion of numerous drugs and endogenous substrates. Patients with Dubin-Johnson syndrome due to mutation in ABCC2 gene have elevated urinary coproporphyrin ratio (UCP I/(I + III)). Here we investigated whether this ratio could serve as a biomarker of MRP2 function. Phenotype-genotype relationships were studied in 74 healthy subjects by measuring individual UCP I/(I + III) ratio obtained on 24-hour urine and by analyzing five common SNPs in ABCC2 gene. The UCP I/(I + III) ratio varied from 14.7% to 46.0% in our population. Subjects with 3972TT genotype had a higher ratio (P = .04) than those carrying the C allele. This higher UCP I/(I + III) ratio was correlated with a higher level of isomer I excretion. This study provides a proof of concept that UCP I/(I + III) ratio can be used as a biomarker of MRP2 function in clinical studies as it provides quantitative information about the in vivo activity of MRP2 in a given patient.
Sigma-Aldrich
SB-MRP2-Sf9, membrane preparation for Vesicular Transport Assays, recombinant, expressed in baculovirus infected Sf9 cells
Sigma-Aldrich
SB-MRP2-Sf9-PREDEASY-ATPase kit, membrane based kit for Human MRP2 ATPase Transport Assays
Sigma-Aldrich
SB-ratMrp2-HEK293, MRP2 rat vesicles
Sigma-Aldrich
SB-MRP2-HEK293