MilliporeSigma
WTAP regulates migration and invasion of cholangiocarcinoma cells.
Journal of gastroenterology (2013-01-29)
Hong-Jae Jo, Hye-Eun Shim, Myoung-Eun Han, Hyun-Jung Kim, Ki-Sun Kim, Sungmin Baek, Kyung-Un Choi, Gi-Yeong Hur, Sae-Ock Oh
ABSTRACT
Wilms' tumor 1-associating protein (WTAP) is a nuclear protein that has been associated with the regulation of proliferation and apoptosis. Although its dynamic expression and physiological functions in vascular cells have been reported, its expression and roles in cholangiocarcinoma cells are poorly characterized. To examine the expression of WTAP in patient tissues, we performed immunohistochemistry. To examine motility of cholangiocarcinoma cells, we employed Boyden chamber, wound healing and Matrigel invasion assays, and a liver xenograft model. Immunohistochemistry in patient tissues showed WTAP overexpression in cholangiocarcinoma tissues and correlation of WTAP expression with metastasis of cholangiocarcinoma cells. Overexpression or knockdown of WTAP significantly increased or decreased the motility of cholangiocarcinoma cells. Moreover, WTAP overexpression or knockdown significantly increased or decreased tumorigenicity of cholangiocarcinoma cells in an orthotopic xenograft model. Furthermore, microarray study showed that WTAP induce the expressions of MMP7, MMP28, cathepsin H and Muc1. WTAP is overexpressed in cholangiocarcinoma and regulates motility of cholangiocarcinoma cells.