MilliporeSigma
  • Catalytic Enantioselective Reaction of α-Aminoacetonitriles Using Chiral Bis(imidazoline) Palladium Catalysts.
Catalytic Enantioselective Reaction of α-Aminoacetonitriles Using Chiral Bis(imidazoline) Palladium Catalysts.
Angewandte Chemie (International ed. in English) (2015-05-28)
Masaru Kondo, Tomoki Nishi, Tsubasa Hatanaka, Yasuhiro Funahashi, Shuichi Nakamura
ABSTRACT
The catalytic enantioselective reaction of diphenylmethylidene-protected α-aminoacetonitriles with imines has been developed. Good yields and diastereo- and enantioselectivities were observed for the reaction of various imines using chiral bis(imidazoline)/Pd catalysts. The reaction of α-aminonitriles with di-tert-butyl azodicarboxylate afforded chiral α,α-diaminonitriles in high yields with high enantioselectivities.
Sigma-Aldrich
Di-tert-butyl azodicarboxylate, purum, ≥98.0% (GC)