MilliporeSigma
  • Sequential molecular analysis of circulating MCAM/MUC18 expression: a promising disease biomarker related to clinical outcome in melanoma.
Sequential molecular analysis of circulating MCAM/MUC18 expression: a promising disease biomarker related to clinical outcome in melanoma.
Archives of dermatological research (2014-06-07)
Maria Cristina Rapanotti, Tara Mayte Suarez Viguria, Gaetana Costanza, Ilaria Ricozzi, Andrea Pierantozzi, Alessandro Di Stefani, Elena Campione, Sergio Bernardini, Sergio Chimenti, Augusto Orlandi, Luca Bianchi
ABSTRACT
MCAM/MUC18 is a cell adhesion molecule associated with higher incidence of relapse in melanoma. The purpose of our study was to evaluate its role as a promising disease biomarker of progression through sequential molecular MCAM/MUC18 RT-PCR assay on serial blood samples collected during the clinical follow-up of 175 melanoma patients in different American Joint Committee on Cancer (AJCC) stages. MCAM/MUC18 molecular detection, found at least once in 22 out of the 175 patients, was significantly associated with poor prognosis and death (p < 0.001), regardless of the AJCC stages. Positive expression, either if primarily present or later acquired, was associated with melanoma progression, whereas patients primarily negative or with subsequent loss gained clinical remission or stable disease, even if in advanced stages (p < 0.005). Six AJCC advanced stages always MCAM/MUC18 negative are in complete remission or with a stable disease (p < 0.007). Semiquantitative immunohistochemical MCAM/MUC18 staining on corresponding primary melanomas was related to peripheral molecular expression. Correlations between circulating molecular and tissutal immunohistochemical detection, primary tumour thickness, AJCC stages and clinical outcome were statistically evaluated using Student's t test, ANOVA, Spearman's rank correlation test, Pearson χ (2)-test and McNemar's test. In our investigation, MCAM/MUC18 expression behaves as a "molecular warning of progression" even in early AJCC patients otherwise in disease-free conditions. Achievement of this molecule predicted the emergence of a clinically apparent status, whereas absence or persistent loss was related to a stable disease or to a disease-free status. If confirmed in larger case series, MCAM/MUC18 molecular expression could predict good or poor clinical outcome, possibly becoming a promising prognostic factor.
Sigma-Aldrich
Anti-MCAM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution