MilliporeSigma
  • Identification of stable S-adenosylmethionine (SAM) analogues derivatised with bioorthogonal tags: effect of ligands on the affinity of the E. coli methionine repressor, MetJ, for its operator DNA.
Identification of stable S-adenosylmethionine (SAM) analogues derivatised with bioorthogonal tags: effect of ligands on the affinity of the E. coli methionine repressor, MetJ, for its operator DNA.
Organic & biomolecular chemistry (2009-02-06)
Catherine Joce, Jamie Caryl, Peter G Stockley, Stuart Warriner, Adam Nelson
ABSTRACT
The efficient synthesis of a range of stable SAM mimetics, and their ability to promote the binding of the E. coli methionine repressor (MetJ) to its operator DNA, is described.
Sigma-Aldrich
Methyl 4-iodobutyrate, 95%