MilliporeSigma
  • A ninhydrin-based assay to quantitate the total protein content of tissue samples.
A ninhydrin-based assay to quantitate the total protein content of tissue samples.
Analytical biochemistry (2001-04-26)
B Starcher
ABSTRACT
Quantitation of small tissue samples for total protein content is essential for many biochemical analyses. In this study a ninhydrin method for measuring the total protein content of tissue hydrolysates is presented. The ninhydrin reagent is stable at room temperature for up to 1 month in the ethylene glycol-sodium acetate solvent system without the requirement for a nitrogen atmosphere. The reaction was very accurate and precise, with intra- and interassay variations of less than 3% when 5 microg of protein was assayed. All proteins that were investigated contributed the same color intensity per microgram protein as bovine serum albumin. This assay was several times more sensitive than the Coomassie reaction and linear over a greater range of protein concentration.
Product Description
Supelco
Ninhydrin reagent according to Stahl, for TLC derivatization