MilliporeSigma
  • Differential expression of annexins I, II and IV in human tissues: an immunohistochemical study.
Differential expression of annexins I, II and IV in human tissues: an immunohistochemical study.
Histochemistry and cell biology (1998-08-28)
R Dreier, K W Schmid, V Gerke, K Riehemann
ABSTRACT
Annexins constitute a family of Ca2+- and phospholipid-binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. To elucidate a possible correlation of those functional proposals with the tissue distribution of annexins, we analysed immunohistochemically the expression of annexins I, II and IV in a broad variety of human tissues. Annexins I and II were chosen for this study since their functionally relevant N-terminal domains are structurally closely related, whilst annexin IV is structurally less related to the former two proteins. The study revealed distinct expression patterns of annexins I, II and IV throughout the body. Annexin I was found in leucocytes of peripheral blood, tissue macrophages and T-lymphocytes and in certain epithelial cells (respiratory and urinary system, superficial cells of non-keratinised squamous epithelium), annexin II in endothelial cells, myoepithelial cells and certain epithelial cells (mainly respiratory and urinary system), whereas annexin IV was almost exclusively found in epithelial cells. Epithelia of the upper respiratory system, Bowman's capsule, urothelial cells, mesothelial cells, peripheral nerves, the choroid plexus, ependymal cells and pia mater and arachnoid of meninges generally strongly expressed all three annexins investigated. The characteristic expression in different tissues and the intracellular distribution indicates that the three annexins investigated are involved in aspects of differentiation and/or physiological functions specific to these tissues.
Sigma-Aldrich
Annexin A1 (MRQ-3) Mouse Monoclonal Antibody