MilliporeSigma
  • The prognostic significance of Ki-67 labeling indices for oligodendrogliomas.
The prognostic significance of Ki-67 labeling indices for oligodendrogliomas.
Neurosurgery (1997-10-08)
S W Coons, P C Johnson, D K Pearl
ABSTRACT
The prognostic significance of quantitative measurement of tumor proliferative activity was evaluated for oligodendroglial tumors. Ki-67/MIB-1 immunochemistry was used to measure proliferative activity in 81 oligodendrogliomas and oligoastrocytomas. The relationship among survival, proliferation, histological features, and clinical variables were evaluated using a Cox proportional hazards analysis. After stratifying by histological grade and adjusting for age at diagnosis, there was a significant association between the Ki-67/MIB-1 labeling index (LI) (percentage of positive cells) and survival (P = 0.04). This association was illustrated further by the significantly different survival of two groups based on LI ranges of less than or equal to 5 and greater than 5 (P < 0.0001). The poor correlation between mitotic figures and survival in oligodendrogliomas that has been reported previously emphasizes the need for an accurate method to measure proliferative activity. Our study demonstrated the usefulness of the Ki-67/MIB-1 LI and demonstrated that LI ranges can be defined for clinical application.
Sigma-Aldrich
Ki-67 (SP6) Rabbit Monoclonal Antibody