MilliporeSigma
  • Tumor-infiltrating Foxp3-CD4+CD25+ T cells predict poor survival in renal cell carcinoma.
Tumor-infiltrating Foxp3-CD4+CD25+ T cells predict poor survival in renal cell carcinoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2007-04-04)
Sameer A Siddiqui, Xavier Frigola, Sandra Bonne-Annee, Maria Mercader, Susan M Kuntz, Amy E Krambeck, Shomik Sengupta, Haidong Dong, John C Cheville, Christine M Lohse, Christopher J Krco, W Scott Webster, Bradley C Leibovich, Michael L Blute, Keith L Knutson, Eugene D Kwon
ABSTRACT
Regulatory T cells (Tregs) have been implicated as inhibitors of antitumoral immunity, and evidence suggests that elimination of Tregs may augment natural and pharmacologic immunity. We tested for the presence of putative Tregs within renal cell carcinoma (RCC) tumors. We identified 170 patients who underwent radical or partial nephrectomy for clear cell RCC between 2000 and 2002. Specimens were stained with anti-CD4, anti-CD25, and anti-Foxp3 antibodies and examined using confocal microscopy. Associations of CD4(+)CD25(+)Foxp3(-) and CD4(+)CD25(+)Foxp3(+) T cells with death from RCC were evaluated using Cox proportional hazards regression models. At last follow-up, 46 of 170 patients had died; of these, 37 died from RCC at a median of 1.4 years following nephrectomy (range, 0-4.4). Among the 124 remaining patients, median follow-up was 3.7 years (range, 0-5.7). Forty-three (25.3%) tumors harbored CD4(+)CD25(+)Foxp3(+) T cells. The presence of Foxp3(+) T cells was not significantly associated with RCC death univariately. One hundred forty-three (84.1%) tumors harbored CD4(+)CD25(+)Foxp3(-) T cells. The indicator for >or=10% CD4(+)CD25(+)Foxp3(-) T cells was significantly associated with RCC death univariately [risk ratio (RR), 2.60; 95% confidence interval (95% CI), 1.35-4.98; P = 0.004], after adjusting for tumor B7-H1 expression (RR, 2.53; 95% CI, 1.32-4.85; P = 0.005) and lymphocytic infiltration (RR, 2.53; 95% CI, 1.32-4.87; P = 0.005). Increased presence of CD4(+)CD25(+)Foxp3(+) T cells was not significantly associated with RCC death. In contrast, CD4(+)CD25(+)Foxp3(-) T cells, which may represent a unique set of Tregs or activated helper T cells, was significantly associated with outcome.
Sigma-Aldrich
CD25 (4C9) Mouse Monoclonal Antibody