MilliporeSigma
  • Immunohistochemical detection of cyclin D1 using optimized conditions is highly specific for mantle cell lymphoma and hairy cell leukemia.
Immunohistochemical detection of cyclin D1 using optimized conditions is highly specific for mantle cell lymphoma and hairy cell leukemia.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2001-01-06)
R N Miranda, R C Briggs, M C Kinney, P A Veno, R D Hammer, J B Cousar
ABSTRACT
Mantle cell lymphoma (MCL) is more aggressive when compared with other lymphomas composed of small, mature B lymphocytes. Cyclin D1 is overexpressed in MCL as a result of the translocation t(11;14)(q13;q32). Cyclin D1 immunohistochemistry in fixed, paraffin-embedded tissue contributes to the precise and reproducible diagnosis of MCL without the requirement of fresh tissue. However, its use in bone marrow biopsies is not well established. In addition, increased levels of cyclin D1 mRNA have been found in hairy cell leukemia but have not consistently been detected by immunohistochemistry. We used a polyclonal antibody and heat-induced antigen retrieval conditions to evaluate 73 fixed, paraffin-embedded bone marrow, spleen, and lymph node specimens with small B-cell infiltrates, obtained from 55 patients. Cyclin D1 was overexpressed in 13/13 specimens of MCL (usually strong, diffuse reactivity in most tumor cells) and in 14/14 specimens of hairy cell leukemia (usually weak, in a subpopulation of tumor cells). No reactivity was detected in five cases of B-chronic lymphocytic leukemia; five cases of splenic marginal zone lymphoma; six cases of nodal marginal zone cell lymphoma; two cases of gastric marginal zone cell lymphoma; or ten benign lymphoid infiltrates in bone marrow, spleen, or lymph nodes. In summary, although the total number of studied cases is small and a larger series of cases may be required to confirm our data, we present optimized immunohistochemical conditions for cyclin D1 in fixed, paraffin-embedded tissue that can be useful in distinguishing MCL and hairy cell leukemia from other small B-cell neoplasms and reactive lymphoid infiltrates.
Sigma-Aldrich
CD11c (5D11) Mouse Monoclonal Antibody