MilliporeSigma
A novel transcription factor, T-bet, directs Th1 lineage commitment.
Cell (2000-04-13)
S J Szabo, S T Kim, G L Costa, X Zhang, C G Fathman, L H Glimcher
ABSTRACT
Naive T helper cells differentiate into two subsets, Th1 and Th2, each with distinct functions and cytokine profiles. Here, we report the isolation of T-bet, a Th1-specific T box transcription factor that controls the expression of the hallmark Th1 cytokine, IFNgamma. T-bet expression correlates with IFNgamma expression in Th1 and NK cells. Ectopic expression of T-bet both transactivates the IFNgamma gene and induces endogenous IFNgamma production. Remarkably, retroviral gene transduction of T-bet into polarized Th2 and Tc2 primary T cells redirects them into Th1 and Tc1 cells, respectively, as evidenced by the simultaneous induction of IFNgamma and repression of IL-4 and IL-5. Thus, T-bet initiates Th1 lineage development from naive Thp cells both by activating Th1 genetic programs and by repressing the opposing Th2 programs.
Sigma-Aldrich
T-bet (MRQ-46) Rabbit Monoclonal Antibody