MilliporeSigma
  • Expression of B-lymphocyte-associated transcription factors in human T-cell neoplasms.
Expression of B-lymphocyte-associated transcription factors in human T-cell neoplasms.
The American journal of pathology (2003-02-25)
Teresa Marafioti, Stefano Ascani, Karen Pulford, Elena Sabattini, Milena Piccioli, Margaret Jones, Pier Luigi Zinzani, Georges Delsol, David Y Mason, Stefano A Pileri
ABSTRACT
In this study we have investigated the expression of three B-cell-associated transcription factors in normal lymphoid tissue and in T-cell neoplasms (three cell lines, and more than 50 biopsy samples). Nuclear OCT-1 immunoreactivity was seen in normal B cells, in many extrafollicular T cells, and in a heterogeneous pattern (ranging in intensity from weak to moderate) in most T-cell neoplasms. OCT-2 immunostaining was primarily restricted in normal lymphoid tissue to B cells, and was absent from most T-cell neoplasms. In contrast, immunostaining for BOB-1/OCA-B--essentially restricted to B cells in normal lymphoid tissue, with the exception of activated T-lymphocytes--was seen in all of the T-cell lines tested and the majority of the tumor cells in all categories of T-cell lymphoma. Thus labeling for each of these three B-cell-associated transcription factors can be seen to varying degrees in T-cell neoplasms. However, the high frequency of BOB-1 expression in T-cell neoplasms, in contrast to its absence from resting peripheral T cells, suggests that its expression might be a prerequisite for neoplastic transformation, and prompts a search for the transcriptional target(s) of this factor in T cells.
Sigma-Aldrich
T-bet (MRQ-46) Rabbit Monoclonal Antibody
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
100 mg
Please contact Customer Service for Availability
€42.20
To order products, please contact your local dealer.