MilliporeSigma
  • Microfluidic oxygen imaging using integrated optical sensor layers and a color camera.
Microfluidic oxygen imaging using integrated optical sensor layers and a color camera.
Lab on a chip (2013-02-28)
Birgit Ungerböck, Verena Charwat, Peter Ertl, Torsten Mayr
ABSTRACT
In this work we present a high resolution oxygen imaging approach, which can be used to study 2D oxygen distribution inside microfluidic environments. The presented setup comprises a fabrication process of microfluidic chips with integrated luminescent sensing films combined with referenced oxygen imaging applying a color CCD-camera. Enhancement of the sensor performance was achieved by applying the principle of light harvesting. This principle enabled ratiometric imaging employing the red and the green channel of a color CCD-camera. The oxygen sensitive emission of platinum(ii)-5,10,15,20-tetrakis-(2,3,4,5,6-pentafluorphenyl)-porphyrin (PtTFPP) was detected by the red channel, while the emission of a reference dye was detected by the green channel. This measurement setup allowed for accurate real-time 2D oxygen imaging with superior quality compared to intensity imaging. The sensor films were subsequently used to measure the respiratory activity of human cell cultures (HeLa carcinoma cells and normal human dermal fibroblasts) in a microfluidic system. The sensor setup is well suited for different applications from spatially and temporally resolving oxygen concentration inside microfluidic channels to parallelization of oxygen measurements and paves the way to novel cell based assays, e.g. in tissue engineering, tumor biology and hypoxia reperfusion phenomena.
Sigma-Aldrich
Fibronectin bovine plasma, powder, BioReagent, suitable for cell culture