MilliporeSigma
  • An easily accessible isoindigo-based polymer for high-performance polymer solar cells.
An easily accessible isoindigo-based polymer for high-performance polymer solar cells.
Journal of the American Chemical Society (2011-08-19)
Ergang Wang, Zaifei Ma, Zhen Zhang, Koen Vandewal, Patrik Henriksson, Olle Inganäs, Fengling Zhang, Mats R Andersson
ABSTRACT
A new, low-band-gap alternating copolymer consisting of terthiophene and isoindigo has been designed and synthesized. Solar cells based on this polymer and PC(71)BM show a power conversion efficiency of 6.3%, which is a record for polymer solar cells based on a polymer with an optical band gap below 1.5 eV. This work demonstrates the great potential of isoindigo moieties as electron-deficient units for building donor-acceptor-type polymers for high-performance polymer solar cells.
Sigma-Aldrich
6,6′-Dibromoisoindigo